Orders LinkContactUsLinkComponent
Media/Lab Bottles, Electron Microscopy Sciences
Media/Lab Bottles, Electron Microscopy Sciences
Catalog # 102094-956
Media/Lab Bottles, Electron Microscopy Sciences
Catalog # 102094-956
Supplier:  
Media/Lab Bottles, Electron Microscopy Sciences
Media/Lab Bottles, Electron Microscopy Sciences
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
250ML AMBER LAB BOTTLE Catalog #89051-808
Product 89051-808 is similar to 102094-956 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Graduated bottles have a large opening, no-drip pour lip and sloping shoulders to facilitate pouring and cleaning.
These bottles are widely used for mixing and storage of culture media, fixation and dehydrating materials. Bottles show approximate volume graduations, and have large frosted writing area. Clear bottles are Wheaton '400' borosilicate glass. Low alkali content helps to prevent changes in pH and maintain purity of contents. Comes with 'PTFE'-lined cap. Autoclavable. Amber bottles are manufactured from '500' borosilicate glass to protect light-sensitive samples.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.