Orders LinkContactUsLinkComponent
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
Tray & Cover for Snap Clip Modules
Catalog # 102094-724
Supplier #:  63484-83
ProPlate™ MicroArray System, Electron Microscopy Sciences
Catalog # 102094-724
Supplier:  
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
Tray & Cover for Snap Clip Modules
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
Specifications
  • Description:
    Tray & Cover for Snap Clip Modules
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.