ProPlate™ MicroArray System, Electron Microscopy Sciences
Product Details & Documents
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
Specifications
- Description:2 Well Module, 16.5x34 mm Well
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)