Orders LinkContactUsLinkComponent
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
2 Well ProPlate Slide Module Tray Set, 3-16.5x34 mm Well, Stainless Steel Spring Clips
  102094-624
Supplier #:  63484-02
ProPlate™ MicroArray System, Electron Microscopy Sciences
  102094-624
 :  
ProPlate™ MicroArray System, Electron Microscopy Sciences
ProPlate™ MicroArray System, Electron Microscopy Sciences
2 Well ProPlate Slide Module Tray Set, 3-16.5x34 mm Well, Stainless Steel Spring Clips
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
  • Description:
    2 Well ProPlate Slide Module Tray Set, 3-16.5x34 mm Well, Stainless Steel Spring Clips
  • Cat. No.:
    102094-624
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)