Floatation Bath & Flotation Workstations, Electron Microscopy Sciences
Product 89207-134 is similar to 102094-532 but manufacturer and specifications may vary.
Product Details & Documents
This Flotation Workstation helps to make your tissue preparation easier, safer and more efficient.
- Magnetic non-contact switch function for years flawlessly
- Microprocessor-controlled heating, ambient to 65°C
- Flat membrane switches for safety and simplicity
- Fluorescent white lighting positioned for improve tissue viewing glare-free
- Heat-resistant plastic housing maintains cosmetic appearance
- Removable glass dish for easy cleaning
- 2 x 6" or 2 x 8" Convection Slide dryer & 2 x 2" HISTO Orientator™ for eliminating wrinkles
The unit comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled electronics and a high contrast background for optimum specimen viewing combined with the HISTO Orientator™ for flattening sections and a Slide Dryer, all in one compact unit. The systems are now available in Low Profile 8 x 8 x 2 ¼" deep dish versions as well as for those labs wanting to float larger ribbons, 8 x 11 x 2".
For those flotation bath applications not requiring the add convenience of Slide Dryer and HISTO Orientator™, the basic Flotation Bath provides all the functional need of the Flotation Workstation at the affordable price. The baths again come in two sizes , 8 x 8 x 2 1/4" or 8 x 11 x 2".
Specifications
- Description:Workstation 2 x 8" Slide Dryer, 220 VAC
- Size:8 x 11 x 2" Dish
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)