True North™ Cool Containers, Electron Microscopy Sciences
Product Details & Documents
These rigid single mold polyurethane containers provides durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra colduse with ice, LN2, ice-salt slurries, and more.
- Convenient lid nests beneath bucket or pan and easily removes with comfortable recessed knobs
- Snug fit minimizes evaporation loss
- Handy pour spout for pouring melted ice, water, and solutions
- Ergonomic handles safely transport bucket or pan from work area to sink
- Deep internal cavity has enough room to add a rack
- Tapered sides make stacking and storage compact
- Available in a bucket or pan and in multiple sizes
Specifications
- Description:Cool Containers, 4L Pan, Artic Blue
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.