Immuno Stain Moisture Chambers, Electron Microscopy Sciences
Product Details & Documents
Innovative, low-cost, high precision moisture chamber for ten slides made by Evergreen.
- Fabricated out of heavy-duty polystyrene
- Air-tight design keeps moisture in
- Stackable design saves counter and refrigerator space
- Available in clear, black and amber
Each moisture chamber is divided into ten individual compartments with approximately half-inch empty space between the compartments. When the chamber lid is closed, eight barrier dividers are placed into the empty spaces between the microscope slide compartments completely isolating all compartments. This is a very desirable feature when doing immuno staining.
The microscope slides are placed on four pedestal posts and four corner posts each 0.460" (11.5 mm) high thus raising the surface of the slides approximately half-inch off the floor to keep the slides away from the water below and to make the slides easily retrievable (either by hand or by forcep).
Specifications
- Color:Clear
- Description:Immuno Stain Moisture Chamber
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.