Orders LinkContactUsLinkComponent
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Assorted colors
Catalog # 102093-618
Supplier #:  61600-A
81-Well Storage Rack, Electron Microscopy Sciences
Catalog # 102093-618
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Electron Microscopy Sciences
81-Well Storage Rack, Assorted colors
Price
$159.15
Log in for availability
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
  • Molded grid lines on the lid
  • Imprinted coordinates on the bottom of base
  • Molded reference point on the lid
  • Rack measures 5.125×5.125×1.75 cm
Specifications
  • Color:
    Assorted colors
  • Description:
    81-Well Storage Rack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)