Orders LinkContactUsLinkComponent
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 2X, 1 l, Bottle
Catalog # 102092-218
Supplier #:  19343-10
Phosphate Buffered Saline, Electron Microscopy Sciences
Catalog # 102092-218
Supplier:  
CAS Number:  
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, 2X, 1 l, Bottle
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Specifications
Specifications
  • Pack type:
    Bottle
  • Description:
    Phosphate Buffered Saline, 2X
  • Size:
    1 l
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)