Phosphate Buffered Saline, Electron Microscopy Sciences
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Specifications
Specifications
- Pack type:Bottle
- Description:Phosphate Buffered Saline, 10X
- Size:1 l
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.