Pop-Up Slide Holder, Electron Microscopy Sciences
Product Details & Documents
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".
Specifications
- Description:30 Slide Pop-Up Slide Folder
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)