$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SEED DIRECT PCR KIT 1000PREP
Catalog # CA11012-532
Supplier:
Omega Bio-Tek
Expand image
SEED DIRECT PCR KIT 1000PREP
Catalog # CA11012-532
Supplier:
Omega Bio-Tek
Supplier Number:
TQ2320-02
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International