$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
DEMONSTRATION SLINKY SPRING
000000000002407477
:
OSAW INDUSTRIAL PRODUCTS SE
DEMONSTRATION SLINKY SPRING
000000000002407477
:
OSAW INDUSTRIAL PRODUCTS SE
:
SP254A
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH