Nunc® Barcode Scanners
Catalog # CA73521-418
Supplier: Thermo Fisher Scientific
Supplier #: 330050
Product Details & Documents
Nunc bar code scanners with plug and play functionality play a critical role in sample tracking
The scanners automatically switch between code 128, code 39, interleave 2 of 5 and several other barcodes. These scanners are durable low-cost, and easy to use.
Documents
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature