Snap Caps, DWK Life Sciences
Natural cap, 11 mm, PTFE/Red Rubber
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 242786
Jump to:
Product Details & Documents
Eliminate the need for crimping and decapping tools.
- Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Documents
Specifications
- Septum:PTFE/Red Rubber
- Description:Natural cap
- Size:11 mm