Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Product Details & Documents
Accurate to within ±0.01 pH at 25 °C (±0.02 for pH 9 and 9.18).
Available as clear or color-coded solutions.
Documents
Certificates
Search by Lot Number0029 Product Specifications 0031 Product Specifications 0097 Product Specifications 0098 Product Specifications 0099 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Specifications
Specifications
- Pack type:Poly bottle
- Color:Yellow
- Size:4 L (1.1 gal.)
- pH:7.0
- pH accuracy:±0.01
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



