Order Entry
Canada
Orders LinkContactUsLinkComponent
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

±0.01, Poly bottle, 4 L (1.1 gal.), 4.01, Red
Catalog # CAMK009710
Supplier #:  MACR0097-10
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Catalog # CAMK009710
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

±0.01, Poly bottle, 4 L (1.1 gal.), 4.01, Red

CAS Number:  
MOC:  Non-Regulated, With Information
Price
$213.73
Log in for availability
Each(1/Itm/Bx) Case of 4 (1/ Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Accurate to within ±0.01 pH at 25 °C (±0.02 for pH 9 and 9.18).

Available as clear or color-coded solutions.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Red
  • Size:
    4 L (1.1 gal.)
  • pH:
    4.01
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.