Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Buffer Solutions

VWR® Buffer Solutions

Acetate, Poly bottle, 4 L, 4.00, Colorless
Catalog # CA71006-844
VWR® Buffer Solutions
Catalog # CA71006-844
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions

Acetate, Poly bottle, 4 L, 4.00, Colorless

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ACETATE BUFFER PH4.0 4L Catalog #BDH7300-4

 

ACETATE BUFFER 1GAL 1GL Catalog #RC501

 

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are for calibrating pH meters

They are traceable to NIST standards.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    4 L
  • pH buffer type:
    Acetate
  • pH:
    4.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.