VWR® Buffer Solutions
Product CAAA40447-K7 is similar to CA71006-708 but manufacturer and specifications may vary.
Product BDH5010-500ML is similar to CA71006-708 but manufacturer and specifications may vary.
Product RC1493-1 is similar to CA71006-708 but manufacturer and specifications may vary.
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.
Documents
Certificates
Search by Lot Numberspec_71006-730 spec_71006-744 spec_71006-768 spec_71006-774 spec_71006-810
Product specification
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)