Orders LinkContactUsLinkComponent
Lighted Forceps, Scissors, Electron Microscopy Sciences
Lighted Forceps, Scissors, Electron Microscopy Sciences
Forceps, 15 cm, Smooth Fine Tip
Catalog # 101764-998
Supplier #:  77905-02
Lighted Forceps, Scissors, Electron Microscopy Sciences
Catalog # 101764-998
Lighted Forceps, Scissors, Electron Microscopy Sciences
Forceps, 15 cm, Smooth Fine Tip
Price
$676.32
Log in for availability
Each Kit of 2
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
See your samples and your embeddings in a whole new light!
Increases the visibility of the specimen
Allows for accurate orientation of tissue and specimen
Minimizers floaters
Reduces embedding and prep time
These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
 : Each kit includes: one instrument, power pack and battery charger.
Specifications
  • Description:
    Forceps
  • Length:
    15 cm
  • Tip style:
    Smooth Fine Tip
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.