Orders LinkContactUsLinkComponent
StatMark Pen, Electron Microscopy Sciences
StatMark Pen, Electron Microscopy Sciences
StatMark Pen
Catalog # 101764-992
Supplier #:  72109-01
StatMark Pen, Electron Microscopy Sciences
Catalog # 101764-992
StatMark Pen, Electron Microscopy Sciences
StatMark Pen, Electron Microscopy Sciences
StatMark Pen
Price
$20.14
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
  • Permanent ink in an ultrafine tip
  • Quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes
  • Do not dry out easily
  • Last twice as long as many of the leading 'laboratory' pens
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.
Documents
Specifications
  • Description:
    StatMark Pen
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.