Orders LinkContactUsLinkComponent
SEM Conductive Tapes, Electron Microscopy Sciences
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape, 50 mm
Catalog # 101764-950
Supplier #:  77817-50-AL
SEM Conductive Tapes, Electron Microscopy Sciences
Catalog # 101764-950
SEM Conductive Tapes, Electron Microscopy Sciences
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape, 50 mm
Price
$589.07
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We now offer a new line of conductive tapes, that are electrically conductive and have a clean background. They were developed especially for scanning electron microscopy.
Documents
Specifications
  • Description:
    Double Sided Carbon Tape, 50 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)