Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PHENOL LOOSE CRYST LB GRD 10KG
Catalog # CA71007-956
Supplier:
VWR
SDS
Expand image
PHENOL LOOSE CRYST LB GRD 10KG
Catalog # CA71007-956
Supplier:
VWR
Supplier Number:
68095-580
SDS
Related Information
Documentation
SDS
ghs_ANAC68095
sds_ANAC68095
Product specification
spec_CA71007-956
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH