Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Buffer Solutions

VWR® Buffer Solutions

Catalog # CA71006-766
VWR® Buffer Solutions
Catalog # CA71006-766
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER PH 7.00 5GAL 5GL Catalog #RC15505

Product RC15505 is similar to CA71006-766 but manufacturer and specifications may vary.

BDH BUFFER REF STD PH7 COLORLESS 20L Catalog #BDH5056-20L

Product BDH5056-20L is similar to CA71006-766 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are for calibrating pH meters

They are traceable to NIST standards.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.