VWR® Buffer Solutions
Product CAJT5608-3 is similar to CA71006-762 but manufacturer and specifications may vary.
Product RC15501 is similar to CA71006-762 but manufacturer and specifications may vary.
Product BDH5048-4L is similar to CA71006-762 but manufacturer and specifications may vary.
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.
Documents
Certificates
Search by Lot Numberspec_71006-730 spec_71006-744 spec_71006-768 spec_71006-774 spec_71006-810
Product specification
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)