Orders LinkContactUsLinkComponent
VWR® Buffer Solutions
VWR® Buffer Solutions
Catalog # CA71006-756
VWR® Buffer Solutions
Catalog # CA71006-756
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BDH BUFFER REF STD PH5 COLORLESS 20L Catalog #BDH5036-20L
Product BDH5036-20L is similar to CA71006-756 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)