Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FBS, ULTIMATE GRD, US ORIGIN, 100ML, GI
Catalog # 97068-094
Supplier:
VWR
SDS
Certificates
Expand image
FBS, ULTIMATE GRD, US ORIGIN, 100ML, GI
Catalog # 97068-094
Supplier:
VWR
Supplier Number:
1600-100G
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_1600
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH