Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Micro Cover Glasses, Rectangular

VWR® Micro Cover Glasses, Rectangular

Cover glasses, rectangular, 40×24 mm, #1
Catalog # 48393-060
Supplier:   VWR International
VWR® Micro Cover Glasses, Rectangular
Catalog # 48393-060
Supplier:  VWR International
VWR® Micro Cover Glasses, Rectangular
VWR® Micro Cover Glasses, Rectangular

Cover glasses, rectangular, 40×24 mm, #1

Price
$49.34
New Customer Promotional Price
Log in for availability
Each(1/Itm/Bx) Ounce Case of 10 (1/ Itm/Bx)
Product Details & Documents

These micro cover glasses are made from the best quality glass, and each set of numbered slides is uniform in size and thickness.

  • Quality glass construction
  • Uniform sizes
  • Two-slide thickness options available

The No. 1 slides are 0.13 to 0.17 mm in thickness. The No. 1¹/₂ slides are 0.16 to 0.19 mm in thickness.

Packaging: Slides are packed 1 oz. per box.

Specifications
  • Numbering:
    #1
  • Description:
    Cover glasses, rectangular
  • L×W:
    40×24 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.