Order Entry
Canada
Orders LinkContactUsLinkComponent
Methyl red

Methyl red

1 g
Catalog # TCM0421-001G
Supplier:   TCI America
Supplier #:  M0421-1G
Methyl red
Catalog # TCM0421-001G
Supplier:  TCI America
CAS Number:  493-52-7
Methyl red

1 g

CAS Number:  493-52-7
Price
$39.30
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Form: Crystal
Color: Deep Red
Melting point (°C): 182
Lambda max.: 410 nm

Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-2'-carboxylic Acid, Acid Red 2

Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.