Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Ammonia solution 50%

Poly bottle, 1 L
Catalog # RC640-32
Corrosif
Santé
Environnement
Supplier:   Ricca Chemical
 :  R0640000-1A
Ammonia solution 50%
Catalog # RC640-32
Supplier:  Ricca Chemical
CAS Number:  1336-21-6
Ammonia solution 50%
Ammonia solution 50%

Poly bottle, 1 L

CAS Number:  1336-21-6
Price
211,99 $
Log in pour la disponibilité
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonymes:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formule:NH₃
  • Numéro MDL:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Ébullition Pt:~ 36 °C
  • Fusion Pt:~ 0 °C
  • Densité:0.9500 g/cm³
  • Température de conservation:Ambient
  • ONU:2672
  • MARC:8,III
Spécifications

Résultats des tests de spécification

  • Appearance:
    Colorless liquid
  • Assay (vs. Sulfuric Acid/Methyl Red):
    48.5 - 51.5 % (v/v)

Spécifications

  • Pack type:
    Poly bottle
  • Size:
    1 L

Spécifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.