Ethylene Oxide Sterilization Indicator Labels
Self-adhesive, nontoxic, chemical process indicators change from purple to green upon exposure to ethylene oxide.
Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.
The color transition is sensitive to time, temperature, and presence of ethylene oxide. The exposure required for a color change is 600mg/L of ethylene oxide for a minimum of 30 minutes at 50–70% RH and 30°C (86°F). At longer exposure times or higher temperatures, the indicators exhibit a brighter shade of green. When exposed, imprinted indicators 19872-032 read “GREEN IS ETO EXPOSED.” Indicators have 24-month shelf life from date of manufacture when stored at room temperature.
- Description:Plain, for Applicator Gun, 1000 Indicators per Roll
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.