Orders LinkContactUsLinkComponent
VWR® Rapid Transfer Buffer, 10X
VWR® Rapid Transfer Buffer, 10X
Rapid Transfer Buffer, 10X, 4 L
Catalog # 97064-314
   Safer Human and Environmental Health
Supplier:   VWR
Supplier #:  N789-4L
VWR® Rapid Transfer Buffer, 10X
Catalog # 97064-314
Supplier:  VWR
   Safer Human and Environmental Health
CAS Number:  
VWR® Rapid Transfer Buffer, 10X
VWR® Rapid Transfer Buffer, 10X
Rapid Transfer Buffer, 10X, 4 L
CAS Number:  
Price
$766.79
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Rapid Transfer Buffer is a simple, single component system for quick and efficient transfer of proteins from SDS-PAGE gels to membranes for Western blotting applications
  • Fast, efficient transfer in 10–20 minutes
  • Utilizes standard transfer equipment
  • Non-hazardous formulation
  • Transfers to PVDF and nitrocellulose from multiple types of gels
Transfer is completed in 10 or 20 minutes using a standard semi-dry or wet transfer apparatus, respectively. Rapid Transfer Buffer is a methanol-free, non-hazardous formulation that is compatible with PVDF and nitrocellulose membranes. It is used for transfer of proteins from standard Laemmli gels, commercial pre-cast gels, and VWR® NEXT GEL™. The transfer efficiency is equivalent to that observed when using a Tris-Glycine-Methanol transfer buffer.
Specifications
Specifications
  • Description:
    Rapid Transfer Buffer, 10X
  • Size:
    4 L
  • Cat. No.:
    97064-314
  • Environmentally Preferable:
    Safer Human and Environmental Health