Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® MOPS Buffer Solution 10X, Biotechnology Grade

VWR® MOPS Buffer Solution 10X, Biotechnology Grade

500 mL
Catalog # 97062-854
Supplier:   VWR
Supplier #:  E526-500ML
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
Catalog # 97062-854
Supplier:  VWR
CAS Number:  
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
VWR® MOPS Buffer Solution 10X, Biotechnology Grade

500 mL

CAS Number:  
Price
$230.30
Log in for availability
Each (500ml)
Product Details & Documents

Optimized for RNA Agarose Electrophoresis

Ideal for Use with RNA Formaldehyde Gels.

Specifications

Specifications

  • Size:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.