Orders LinkContactUsLinkComponent
Sampling Tubes and Systems, Drager
Silica gel tubes, Type NIOSH
  13453-472
Sampling Tubes and Systems, Drager
  13453-472
 :  
Sampling Tubes and Systems, Drager
Sampling Tubes and Systems, Drager
Silica gel tubes, Type NIOSH
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
  • Description:
    Silica gel tubes, Type NIOSH