Order Entry
Canada
Orders LinkContactUsLinkComponent
 

D-(-)-Ribose ≥98.0%

25 g
  TCR0025-025G
 :  R0025-25G
D-(-)-Ribose ≥98.0%
  TCR0025-025G
 :  50-69-1
D-(-)-Ribose ≥98.0%

25 g

 :  50-69-1
 
$28.10
   
Each (25g)
 
   
 

CAS Number: 50-69-1
MDL Number: MFCD00135453
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Specific rotation [a]20/D: -21 deg (C=1, H2O)

  •  :50-69-1
  •  :C₅H₁₀O₅
  •  :MFCD00135453
  •  :150.13 g/mol
  •  :12,08371
  •  :331 °C (1013 hPa)
  •  :90…95 °C
  •  :0.8 g/cm³ (20 °C)
  •  :Ambient
  •  :200-059-4
 

 

  • Purity/Analysis Method:
    98.0% (GC)

 

  • Size:
    25 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.