Order Entry
Canada
Orders LinkContactUsLinkComponent
Dioctadecyl ether ≥95.0%

Dioctadecyl ether ≥95.0%

25 g
Catalog # TCO0016-025G
Supplier:   TCI America
Supplier #:  O0016-25G
Dioctadecyl ether ≥95.0%
Catalog # TCO0016-025G
Supplier:  TCI America
CAS Number:  6297-03-6
Dioctadecyl ether ≥95.0%

25 g

CAS Number:  6297-03-6
Price
$191.00
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 6297-03-6
MDL Number: MFCD00048498
Molecular Formula: C36H74O
Molecular Weight: 522.99
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 131
Melting point (°C): 63

Other Product Names / Synonyms: Distearyl Ether, Octadecyl Ether

  • CAS Number:6297-03-6
  • Formula:C₃₆H₇₄O
  • MDL Number:MFCD00048498
  • Boiling Pt:131 °C
  • Melting Pt:63 °C

Documents

Specifications

Specification Test Results

  • Purity/Analysis Method:
    95.0% (GC)

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.