Order Entry
Canada
Orders LinkContactUsLinkComponent
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.0%

γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.0%

25 g
Catalog # TCM0707-025G
Supplier:   TCI America
Supplier #:  M0707-25G
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.0%
Catalog # TCM0707-025G
Supplier:  TCI America
CAS Number:  1132-61-2
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.0%

25 g

CAS Number:  1132-61-2
Price
29,50 $
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

[Good's buffer component for biological research]
CAS Number: 1132-61-2
MDL Number: MFCD00006183
Molecular Formula: C7H15NO4S
Molecular Weight: 209.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: MOPS

Synonyms:  3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
  • CAS Number:1132-61-2
  • Formula:C₇H₁₅NO₄S
  • MDL Number:MFCD00006183
  • Molecular Weight:209.27 g/mol
  • Merck Index:13,06288
  • Melting Pt:> 250 °C
  • Flash Pt:110 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:214-478-5

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    98.0% (T)

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.