Universal Reverse Primer, Agilent Technologies
Universal reverse primer, 50
Catalog # 97066-434
Supplier #: 600037
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PRIMER REVERSE UNIVERSAL 200UL 50 RXN Catalog #CA97066-434
Product CA97066-434 is a replacement for 97066-434 and has the same packaging and specifications, but may be a different quantity.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The universal reverse primer serves as the downstream primer in the qPCR reaction by annealing to a universal sequence tag that is added to the cDNA during the reverse transcription protocol.
Applications: PCR
Number of Reactions: 3.125 μM
Physical Form: Liquid
Reactions per Kit: 300
Volume: 200 µL
Documents
Agilent Technologies qPCR Kit Product List
Product Literature
Specifications
Specifications
- Description:Universal reverse primer
- No. of reactions:50
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



