Orders LinkContactUsLinkComponent
VWR® Rapidblock™ Blocking Solution, 10X
VWR® Rapidblock™ Blocking Solution, 10X
100 mL
Catalog # 97064-124
Supplier:   VWR
Supplier #:  M325-100ML
VWR® Rapidblock™ Blocking Solution, 10X
Catalog # 97064-124
Supplier:  VWR
CAS Number:  
VWR® Rapidblock™ Blocking Solution, 10X
VWR® Rapidblock™ Blocking Solution, 10X
100 mL
CAS Number:  
Price
$464.96
Log in for availability
Each
Product Details & Documents
Solution reduces blocking time to 5 minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes.
  • Only 5 Minutes for PVDF or Nitrocellulose Blocking
  • Reduce Background and Cross-Reactivity
  • Enhance Chemiluminescent Signals
  • Protein-Free Formulation
This protein-free formulation minimizes cross-reactivity and non-specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio. Results meet or exceed those obtained with buffers containing dried milk or BSA, which both require 1 hour of blocking time.
Specifications
Specifications
  • Size:
    100 mL