Order Entry
Canada
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol ≥96%, high purity

D-(-)-Sorbitol ≥96%, high purity

Catalog # 97062-202
D-(-)-Sorbitol ≥96%, high purity
Catalog # 97062-202
Supplier:  
CAS Number:  50-70-4
D-(-)-Sorbitol ≥96%, high purity
D-(-)-Sorbitol ≥96%, high purity
CAS Number:  50-70-4
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
D-SORBITOL Catalog #TCS0065-25G

Product TCS0065-25G is similar to 97062-202 but manufacturer and specifications may vary.

D-SORBITOL HIGH PURITY GRADE 500G Catalog #97062-204

Product 97062-204 is similar to 97062-202 but manufacturer and specifications may vary.

Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:98…100 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specification Test Results

  • Arsenic:
    0.0003 %
  • Chloride:
    0.005 %
  • Heavy Metals:
    0.001 %
  • Moisture (KF):
    = 1 %
  • Purity (Anhydrous):
    = 96 %
  • Residue after Ignition:
    0.1 %
  • Sulfate:
    0.01 %
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.