Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, NIST/DIN

pH Buffer Solutions, NIST/DIN

NIST/DIN buffer solution, shelf life: 12 months, Bottle, 250 ml, 10.012
Catalog # 11278-348
Supplier:   Mettler Toledo
Supplier #:  51350058
pH Buffer Solutions, NIST/DIN
Catalog # 11278-348
Supplier:  Mettler Toledo
CAS Number:  
pH Buffer Solutions, NIST/DIN
pH Buffer Solutions, NIST/DIN

NIST/DIN buffer solution, shelf life: 12 months, Bottle, 250 ml, 10.012

CAS Number:  
Price
$82.00
Log in for availability
Each (250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, for example 9.180, offering the highest possible pH precision.

  • Used for calibration/verification purposes
  • Uncertainty: ±0.015 pH

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

Single-use sachets, filled under controlled conditions, guarantee a fresh and uncontaminated solution for every calibration. The round and large opening of the sachets allows for easy sensor insertion and makes calibration a quick and simple task.

Certifications: NIST traceable.

Packaging: Packed in either sachets or bottles.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    NIST/DIN buffer solution, shelf life: 12 months
  • Size:
    250 ml
  • pH:
    10.012

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.