Order Entry
Canada
Orders LinkContactUsLinkComponent
 

3,3,5-Trimethylcyclohexanone ≥98.0%

500 ml
Catalog # TCT0602-500ML
Supplier:   TCI America
 :  T0602-500ML
3,3,5-Trimethylcyclohexanone ≥98.0%
Catalog # TCT0602-500ML
Supplier:  TCI America
CAS Number:  873-94-9
3,3,5-Trimethylcyclohexanone ≥98.0%

500 ml

CAS Number:  873-94-9
Price
$75.70
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 873-94-9
MDL Number: MFCD00019466
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Flash Point (°C): 68
Specific Gravity (20/20): 0.89

  • CAS Number:873-94-9
  • Formula:C₉H₁₆O
  • MDL Number:MFCD00019466
  • Molecular Weight:140.23 g/mol
  • Boiling Pt:190 °C
  • Melting Pt:–10 °C
  • Density:0.89 (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:212-855-9
Specifications

Specification Test Results

  • Purity/Analysis Method:
    98.0% (GC)

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.