Thymolphthalein
Product Details & Documents
CAS Number: 125-20-2
MDL Number: MFCD00005909
Molecular Formula: C28H30O4
Molecular Weight: 430.54
Form: Crystal
Color: White
Melting point (°C): 253
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430.54 g/mol
- Merck Index:12,09542
- Melting Pt:253 °C
- Storage Temperature:Ambient
- EINECS:204-729-7
Documents
Specifications
Specifications
- Size:1 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



