Order Entry
Canada
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol ≥97.0%

D-(-)-Sorbitol ≥97.0%

25 g
Catalog # TCS0065-25G
Supplier:   TCI America
Supplier #:  S0065-25G
D-(-)-Sorbitol ≥97.0%
Catalog # TCS0065-25G
Supplier:  TCI America
CAS Number:  50-70-4
D-(-)-Sorbitol ≥97.0%

25 g

CAS Number:  50-70-4
Price
$29.50
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 50-70-4
MDL Number: MFCD00004708
Molecular Formula: C6H14O6
Molecular Weight: 182.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 295
Melting point (°C): 100
Specific rotation [a]20/D: 106 deg (C=0.4, Acidified (NH4)6Mo7O24)

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C
  • Melting Pt:100 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5

Documents

Specifications

Specification Test Results

  • Purity/Analysis Method:
    97.0% (GC)

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.