VWR® Rapidblock™ Blocking Solution, 10X
Product 97064-124 is a replacement for 97063-124 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Solution reduces blocking time to 5 minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes.
- Only 5 Minutes for PVDF or Nitrocellulose Blocking
- Reduce Background and Cross-Reactivity
- Enhance Chemiluminescent Signals
- Protein-Free Formulation
This protein-free formulation minimizes cross-reactivity and non-specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio. Results meet or exceed those obtained with buffers containing dried milk or BSA, which both require 1 hour of blocking time.
Documents
Specifications
Specifications
- Size:15 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)