VWR® SSC Buffer
Product 97062-390 is similar to 97062-382 but manufacturer and specifications may vary.
Product Details & Documents
SSC Buffer is a hybridization buffer for Northern and Southern blots
VWR® offers SSC in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 ml of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.
Documents
Specifications
Specifications
- Description:SSC BUFFER, 20X READY-PACK™
- Size:2 pack
- Application:Each Ready-Pack™ prepares 1L of 20X concentrate.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)