Orders LinkContactUsLinkComponent
Staticide® Concentrate, ACL Staticide
Staticide® Concentrate, ACL Staticide
Gallon, 3.8 L (1 gal.)
Catalog # 58611-382
Supplier:   ACL Staticide
Supplier #:  3000G
Staticide® Concentrate, ACL Staticide
Catalog # 58611-382
Supplier:  ACL Staticide
Staticide® Concentrate, ACL Staticide
Staticide® Concentrate, ACL Staticide
Gallon, 3.8 L (1 gal.)
Price
$2,125.89
Log in for availability
Each (1US Gal)
Minimum order quantity is 1
Product Details & Documents
Staticide® Concentrate is available to save freight, handling, and storage costs while offering greater flexibility in dilution.
Staticide® Concentrate can be diluted with tap waater, Isopropyl Alcohol, Carbon Tetrachloride, or Perchloroethylene to meet various processing needs.
Staticide® Concentrate should be mixed with water or other solvents in the following concentrate solvent ratios to obtain these formulations: General Purpose Staticide® 1:99, Heavy Duty Staticide® 1:39
Certifications: When properly applied, Staticide® complies with or exceeds all electrostatic decay criteria established for antistatic performance as set forth by NFPA and MIL B-81705, when tested in accordance with Federal Test Std 101B, Method 4046.
Ordering information: Staticide® Concentrate is available in quart, gallon, and 50 gallon drum containers.
Documents
Specifications
  • Description:
    Gallon
  • Size:
    3.8 L (1 gal.)