Jump to:
Product Details & Documents
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
- Description:Simultaneous test set II, for inorganic fumes (sulfur dioxide, chlorine, hydrogen sulfide, hydrogen phosphide, carbonyl chloride)
- Response time:40 s
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extension Hose
Product Information
- Catalog #AvailabilityPrice
- Adapter for Simultaneous test sets, for connection to a bellows pumpAdapter for Simultaneous test sets, for connection to a bellows pumpCA34330-044Log in for availabilityEach - $595.27