2',4',6'-Trihydroxyacetophenone monohydrate ≥98.0%
Product Details & Documents
CAS Number: 480-66-0
MDL Number: MFCD00002287
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 215
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2-Acetylphloroglucinol, Phloroacetophenone
- CAS Number:207-556-5
- Formula:C₈H₈O₄·1H₂O
- MDL Number:MFCD00149091
- Molecular Weight:186.16 g/mol
- Melting Pt:215 °C
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Purity/Analysis Method:98.0% (GC)
Specifications
- Size:5 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



