Orders LinkContactUsLinkComponent
VWR® RNase Removing Solution, Nucleaseliminator
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™ Wipes, 25 pack
Catalog # 97063-074
Supplier:   VWR
Supplier #:  E891-25PK
VWR® RNase Removing Solution, Nucleaseliminator
Catalog # 97063-074
Supplier:  VWR
CAS Number:  
VWR® RNase Removing Solution, Nucleaseliminator
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™ Wipes, 25 pack
CAS Number:  
Price
$147.94
Log in for availability
Each
Product Details & Documents
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips and electrophoresis units. NucleasEliminator™ is supplied as a ready-to-use liquid. A 100 ml bottle will treat up to 100 microcentrifuge tubes or 25 agarose electrophoresis casting trays.
Specifications
Specifications
  • Description:
    NUCLEASELIMINATOR™ Wipes
  • Size:
    25 pack
Specifications